Learn More
Thermo Scientific Chemicals Orexin B, Human
For cellular and molecular biology applications
Supplier: Thermo Scientific Chemicals J65110MCR
Description
Orexin is a neuropeptide that modulates the energy and metabolism balance. The effects of orexin are mediated by one of the two G protein-coupled receptors, orexin receptor 1 or 2. OXRs are expressed in oxinergic projection areas of the brain and regulate food intake by reducing systemic and brain signals that drive over-eating. Synthetic peptide corresponding to the C-terminus of human orexin receptor 1, conjugated to KLH. The immunizing peptide has 70% homology with the rat and mouse gene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Human | |
-30°C to -10°C | |
2899.33 | |
1 mg | |
Light sensitive |
HPLC | |
Lyophilized Powder | |
Orexin B, human | |
RUO - research use only | |
Arg-Ser-Gly-Pro-Pro-Gly-Leu-Gln-Gly-Arg-Leu-Gln-Arg-Leu-Leu-Gln-Ala-Ser-Gly-Asn-His-Ala-Ala-Gly-Ile-Leu-Thr-Met-NH2 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only