Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Orexin Receptor 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533839
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Dog, Horse, Pig (100%) Elephant, Bovine (94%) Marmoset, Panda (88%) Bat (81%).
The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein selectively binds the hypothalamic neuropeptide orexin A. A related gene (HCRTR2) encodes a G-protein coupled receptor that binds orexin A and orexin B.
Specifications
Orexin Receptor 1 | |
Polyclonal | |
Unconjugated | |
HCRTR1 | |
Hcrt receptor 1; Hcrtr1; Hctr1; hypocretin (orexin) receptor 1; hypocretin receptor 1; hypocretin receptor type 1; hypocretin receptor-1; Orexin a receptor; orexin receptor 1; orexin receptor type 1; orexin receptor-1; Orexin-1 receptor; OX1 receptor; OX1R; ox-1-R; ox1-R; OX-R1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100056164, 100855896, 3061, 387287 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O43613, O97661 | |
HCRTR1 | |
Synthetic 16 amino acid peptide from C-terminus of human HCRTR1 / Orexin Receptor 1. | |
50 μg | |
Primary | |
Human, Canine, Monkey, Pig, Horse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction