Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORF73/HHV8 Mouse anti-Nonspecies specific, Clone: HHV8/3606, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30780910UG

 View more versions of this product

Catalog No. NB123091


Add to cart

Description

Description

ORF73/HHV8 Monoclonal specifically detects ORF73/HHV8 in Non-species specific samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

ORF73/HHV8
HHV8/3606
Unconjugated
10 mM PBS with 0.05% BSA
HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV
Recombinant protein corresponding to the latent nuclear antigen 1 molecule of ORF73/HHV8
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Nonspecies specific
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.2 mg/ml
ELISA 1-5 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin
Purified
Mouse
IgG1 Kappa
100 μg
Adaptive Immunity, Immunology, Virology Bacteria and Parasites
Primary
935114
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit