Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORF73/HHV8 Mouse anti-Human, Virus, Janelia Fluor 549, Clone: 4C11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP130176JF549

Catalog No. NBP13176549


Add to cart

Description

Description

ORF73/HHV8 Monoclonal antibody specifically detects ORF73/HHV8 in Human, Virus samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

ORF73/HHV8
4C11
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
HHV8GP78
Recombinant human ORF73/HHV8 (122-329aa) purified from E. coli (AAB62657).
Protein G purified
Adaptive Immunity, Immunology, Virology Bacteria and Parasites
Primary
935114
Human, Virus
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV
Mouse
IgG2a Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human ORF73/HHV8 protein.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit