Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ORF73/HHV8 Rabbit anti-Nonspecies specific, Clone: HHV8/3633R, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30727920UG
Description
ORF73/HHV8 Monoclonal specifically detects ORF73/HHV8 in Non-species specific samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ORF73/HHV8 | |
Monoclonal | |
0.2 mg/ml | |
ELISA 1-5 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin | |
HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV | |
Recombinant protein corresponding to the latent nuclear antigen 1 molecule of ORF73/HHV8 | |
20 μg | |
Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
935114 | |
Store at 4°C. | |
IgG |
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
HHV8/3633R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Non-species specific | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction