Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORF73/HHV8 Rabbit anti-Nonspecies specific, Clone: HHV8/3633R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen ORF73/HHV8
Clone HHV8/3633R
Concentration 0.2 mg/ml
Dilution ELISA 1-5 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122031
View Documents
Novus Biologicals
NBP30727910UG
100 μg
Each for $406.52
Add to cart
 
NB122032
View Documents
Novus Biologicals
NBP30727920UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

ORF73/HHV8 Monoclonal antibody specifically detects ORF73/HHV8 in Nonspecies specific samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

ORF73/HHV8
0.2 mg/ml
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Purified
RUO
Nonspecies specific
HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV
Recombinant protein corresponding to the latent nuclear antigen 1 molecule of ORF73/HHV8
Monoclonal
Protein A or G purified
HHV8/3633R
ELISA 1-5 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin
Unconjugated
Rabbit
Adaptive Immunity, Immunology, Virology Bacteria and Parasites
10 mM PBS with 0.05% BSA
935114
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit