Tocris™ Org 12962 hydrochloride

Selective 5-HT2C agonist

Manufacturer: Tocris™ 3585/50

 View more versions of this product

Catalog No. 35-855-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors Org 12962 hydrochloride
    Purity >99
    Molecular Weight 302.12
    Quantity 50mg
    Product Type Org 12962 hydrochloride

    Selective 5-HT2C receptor agonist (pEC50 values are 7.01, 6.38 and 6.28 for 5-HT2C, 5-HT2A and 5-HT2B respectively). Displays antiaversive effects in a rat model of panic-like anxiety.

    • Chemical Name: 1-[6-Chloro-5-trifluoromethyl)-2-pyridinyl]-piperazine hydrochloride
    • Formula: C10H11ClF3N3.HCl
    • CAS Number: 210821-63-9
    • Solubility: Soluble to 50mM in water and to 100mM in DMSO
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit