Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Org 20599, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Positive allosteric modulator and at higher concentrations direct agonist of GABAA receptors (EC50 = 1.1 μM). Also displays positive modulation of glycine receptors (EC50 = 22.9 μM). Anesthetic steroid; induces hypnosis and loss of righting reflexes in mice.
Specifications
Quantity | 10 mg |
Formula Weight | 438.04 |
Percent Purity | >98% |
Chemical Name or Material | Org 20599 |
Safety and Handling
Recommended Storage : Store at Room Temperature
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title