Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Org 20599, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
156685-94-8
C25H40ClNO3
438.049
NZFNABGZEQPYBX-WIWDAOHRSA-N
(2β, 3α, 5α)-21-Chloro-3-hydroxy-2-(4-morpholinyl)pregnan-20-one
122173107
2-chloro-1-[(2S,3R,5S,8R,9R,10S,13S,14R,17R)-3-hydroxy-10,13-dimethyl-2-morpholin-4-yl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone
CC12CCC3C(C1CCC2C(=O)CCl)CCC4C3(CC(C(C4)O)N5CCOCC5)C
This item is not returnable. View return policy
156685-94-8
C25H40ClNO3
438.049
NZFNABGZEQPYBX-WIWDAOHRSA-N
(2β, 3α, 5α)-21-Chloro-3-hydroxy-2-(4-morpholinyl)pregnan-20-one
122173107
2-chloro-1-[(2S,3R,5S,8R,9R,10S,13S,14R,17R)-3-hydroxy-10,13-dimethyl-2-morpholin-4-yl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone
CC12CCC3C(C1CCC2C(=O)CCl)CCC4C3(CC(C(C4)O)N5CCOCC5)C

Positive allosteric modulator and direct agonist of GABAA receptors

Positive allosteric modulator and at higher concentrations direct agonist of GABAA receptors (EC50 = 1.1 μM). Also displays positive modulation of glycine receptors (EC50 = 22.9 μM). Anesthetic steroid; induces hypnosis and loss of righting reflexes in mice.
Quantity 50 mg
Formula Weight 438.04
Percent Purity >98%
Chemical Name or Material Org 20599

Recommended Storage : Store at Room Temperature

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit