Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Org 24598, >98%, Tocris Bioscience™
GREENER_CHOICE

Catalog No. p-7121266
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibitor of GlyT1; derivative of Org 24598 (Cat. No. 4035).

Potent and selective inhibitor of the glial glycine transporter GlyT1b (IC50 = 6.9 nM). Displays negligible activity at GlyT-2, adrenoreceptors, dopamine, 5HT receptors and noradrenaline, dopamine, 5HT and GABA transporters (pIC50 < 4). Active in vivo.

Chemical Identifiers

CAS 372198-97-5
Molecular Formula C19H20F3NO3
Molecular Weight (g/mol) 367.368
InChI Key KZWQAWBTWNPFPW-QGZVFWFLSA-N
Synonym 2-methyl-3r-3-phenyl-3-4-trifluoromethyl phenoxy propyl amino acetic acid, d00lnk, n-methyl-n-3r-3-phenyl-3-4-trifluoromethyl phenoxy propyl-glycine, methyl-r-3-phenyl-3-4-trifluoromethyl-phenoxy-propyl-amino-acetic acid
PubChem CID 5311285
IUPAC Name 2-[methyl-[(3R)-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]amino]acetic acid
SMILES CN(CCC(C1=CC=CC=C1)OC2=CC=C(C=C2)C(F)(F)F)CC(=O)O

Specifications

Linear Formula C19H20F3NO3
Quantity 10 mg
Solubility Information Soluble to 10mM in water with gentle warming and to 100mM in DMSO
Formula Weight 367.36
Chemical Name or Material N-Methyl-N-[(3R)-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]-glycine
Grade >98%
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.