Learn More
Org 24598 lithium salt, Tocris™
Selective GlyT1 inhibitor
Manufacturer:  R&D Systems 4035/10
Quantity 10mg
 View more versions of this product
Catalog No. 40-351-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
373.3
Org 24598 lithium salt
C19H19F3LiNO3
N-Methyl-N-[(3R)-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]-glycine lithium salt
Specifications
722456-08-8
C19H19F3LiNO3
N-Methyl-N-[(3R)-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]-glycine lithium salt
Org 24598 lithium salt
>98%
373.3
10mg
Safety and Handling
Recommended Storage : Desiccate at Room Temperature