Learn More
Apexbio Technology LLC Org 27569, 10mg. Cas: 868273-06-7 MFCD: MFCD11045300. Cannabinoid CB1 receptor allosteric modulator
SDP
Supplier:  Apexbio Technology LLC B142610
Encompass_Preferred
Org 27569 is an allosteric ligand of the cannabinoid receptor CB1[1].Org 27569 has been reported to promote the agonist CP55940 binding to purified bimane-labeled CB1 receptor and act directly on the CB1 receptor, yet at the same time inhibit receptor function. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5233
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.