Learn More
Eisco™ Organ Pipe Products for Science Education
The EISCO Organ Pipe is a pipe with a movable piston that is marked with the chromatic scale from C (512) to (1024)
Manufacturer:  Eisco™ PH0759
Catalog No. S27966
Add to cart
Description
Description
Specifications
Specifications
Organ Pipe
Movable piston marked with the chromatic scale from C (512) to (1024)
Polished wood