Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Organic Carbon Standard, 1mL = 0.5mg C, 500ppm C, Ricca Chemical

Click to view available options
Quantity:
500mL
Packaging:
Glass amber
877-24-7,7664-38-2,7732-18-5
C8H5KO4
204.222
IWZKICVEHNUQTL-UHFFFAOYSA-M
23676735
potassium;2-carboxybenzoate
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
877-24-7,7664-38-2,7732-18-5
C8H5KO4
204.222
IWZKICVEHNUQTL-UHFFFAOYSA-M
23676735
potassium;2-carboxybenzoate
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
CAS Min % 97.0,0.46,0.1
CAS Max % 100.0,0.48,0.11
Physical Form Liquid
Packaging Glass amber
Concentration or Composition Notes 500ppm C
Equivalence 1 mL = 0.5 mg C
Color Colorless
Quantity 500mL
Chemical Name or Material Organic Carbon Standard