Learn More
Organometallic Oil Standard, 12-Element, SPEX CertiPrep
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$166.00 - $256.00
Specifications
Chemical Name or Material | 12 Element Oil Standard |
---|---|
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation |
Certificate of Analysis | With Certificate of Analysis. |
Grade | Oil |
Solvent or Matrix | Base Oil 75 |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
S12100Y
|
SPEX CertiPrep![]() S12100Y |
100g | Triple-leached LDPE bottle |
Each for $256.00
|
|
S12100Z
|
SPEX CertiPrep![]() S12100Z |
50g | Triple-leached LDPE bottle |
Each for $166.00
|
|
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ag | 100µg/g |
Al | 100µg/g |
Cr | 100µg/g |
Cu | 100µg/g |
Fe | 100µg/g |
Mg | 100µg/g |
Na | 100µg/g |
Ni | 100µg/g |
Pb | 100µg/g |
Si | 100µg/g |
Sn | 100µg/g |
Ti | 100µg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
12 Element Oil Standard | |
With Certificate of Analysis. | |
Base Oil 75 |
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation | |
Oil |
Safety and Handling
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.