Learn More
Selleck Chemical LLCSupplier Diversity Partner Oridonin 50mg 28957-04-2 Isodonol, Rubescenin, NSC-250682  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S233550MG
Oridonin (Isodonol, Rubescenin, NSC-250682), a diterpenoid purified from Rabdosia rubescens, is a traditional agent with antitumor, anti-bacterial and anti-inflammatory effects. Oridonin inhibits AKT1 and AKT2 kinase activity with IC50 of 8.4 μM and 8.9 μM, respectively.
Catalog No. 50-668-0