Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Original Cable Lockout

Gain control of difficult lockout situations. The patented Brady™ Original Cable Lockout is ideal for difficult lockout situations such as large gate valves or oversized devices.

Manufacturer:  Brady™ 45192

 View more versions of this product

Catalog No. 19-103-935


  Check Availability
Add to cart

Specifications

Description

  • Brady's original, patented cable lockout device is perfect for your difficult lockout situations such as large gate valves or oversized devices
  • Simply squeeze device to tighten cable and insert your padlock to secure the handle
  • Body is made of impact modified nylon, available in red, blue, yellow or green
  • Standard lockout device comes with 6 ft. x 1/8 in. diameter cable
  • Additional 6 ft., 8 ft. and 10 ft. cables are sold separately
Description & Specifications

Specifications

Pull-tight
Galvanized steel cable
Impact modified nylon
Blue
1.1 in. x 3 in. x 6.5 in.
Includes 6 ft. galvanized steel able
1/Each
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit