Brady™ Original Cable Lockout

Gain control of difficult lockout situations. The patented Brady™ Original Cable Lockout is ideal for difficult lockout situations such as large gate valves or oversized devices.

Manufacturer: Brady™ 45191

 View more versions of this product

Catalog No. 19-103-934

  Check Availability
Add to Cart

    Specifications

    Dimensions (L x W x H) 1.1 in. x 3 in. x 6.5 in.
    For Use With (Equipment) Galvanized steel cable
    Includes Includes 6 ft. galvanized steel able
    Material Impact modified nylon
    Quantity 1/Each
    Color Yellow
    Type Pull-tight

    • Brady's original, patented cable lockout device is perfect for your difficult lockout situations such as large gate valves or oversized devices
    • Simply squeeze device to tighten cable and insert your padlock to secure the handle
    • Body is made of impact modified nylon, available in red, blue, yellow or green
    • Standard lockout device comes with 6 ft. x 1/8 in. diameter cable
    • Additional 6 ft., 8 ft. and 10 ft. cables are sold separately

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit