Learn More
Thermo Scientific™ Orion™ 136S Advanced Waterproof Portable Conductivity Meter Available on GSA/VA Contract for Federal Government customers only.
Measure and log conductivity, salinity, total dissolved solids (TDS) and temperature using the Thermo Scientific™ Orion™ 136S Intrinsically Safe (US) Portable Conductivity Meter
Manufacturer:  Thermo Scientific™ 0136S3
 View more versions of this product
Catalog No. 13-302-113
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Measure conductivity, salinity, total dissolved solids (TDS) and temperature in environments where non-sparking equipment is required using the Thermo Scientific™ Orion™ 136S Intrinsically Safe (US) Portable Conductivity Meter. Log up to 100 data points and use the RS232 port to transfer data to an external device. Intrinsically safe approval rating by Factory Mutual (FM) for US only.
Features:
  • Temperature compensation options, including automatic or manual linear function and nonlinear function for ultrapure or natural water
  • Calibration via cell constant adjustment method or Thermo Scientific™ AUTO-CAL™, which uses direct entry of conductivity standards
  • Custom LCD
  • 100-point datalogger documents readings for GLP requirements
  • RS-232 interface and software program allow simple data collection and transfer
  • Real time clock and calibration printout
  • Factory Mutual (FM)-approved intrinsically safe, non-sparking meter
Includes:
Meter Only option includes meter, built-in quiver, neckstrap, RS-232 cable and data transfer software.
Meter/Conductivity Cell option includes meter, cell, built-in quiver, neckstrap, RS-232 cable and data transfer software.
Meter Field Kit option includes meter, cell, built-in quiver, neckstrap, RS-232 cable and data transfer software, beakers and standards
Description & Specifications
Specifications
Orion Intrinsically Safe 136S conductivity/TDS/salinity portable meter kit with sensor, field case, beakers and solutions
Orion Intrinsically Safe 136S Conductivity/TDS/Salinity/Temperature Portable Meter
Orion 136S conductivity meter, 013010A Orion DuraProbe 4-cell (K=0.475) conductivity sensor with 3 meter cable, field case, beakers, conductivity standards, 3 AA batteries
0.01 µS up to 99.99 µS; 0.1 µS above 99.99 µS
0 to 1999mg/L
Linear (0.40 to 1.00)
0.1ppt
Linear, nLF (non-linear pure or natural water)
Conductivity
-20° to 120 °C, -4° to 248° F
±0.3°C
1
None
Conductivity, TDS or salinity with temperature
100 with time and date stamp
English
RS232
3 AA batteries (included)
Three AA (included)
CE, FM (IS, Class I Div 1 Group A, B, C, D, T4), Ex (Intrinsically Safe)
-10° to 55°C (ambient)
1.4 lb.
0.010 to 199.9cm-1
3 year meter warranty
Orion Intrinsically Safe 136S conductivity/TDS/salinity portable meter kit with sensor, field case, beakers and solutions
0.1 µS/cm to 500 mS/cm (K=1.0, 0.55 or 0.475) 0.01 µS/cm to 199.9 µS/cm (K=0.1)
0.5% of reading ±1 digit < 500 mS; 1% of reading ±1 digit = < 500 mS
1mg/L
0.0 to 45.0 ppt
2-cell or 4-cell conductivity sensors with built-in temperature
25°C
1
0.1°C
Automatic with ATC temperature probe
LCD
Measure, cal, print, scroll up, scroll down, STO, RCL, clock, power keys
Transfer single or all data points to printer or computer
Automatic by interval or event or manual data logging
8 pin Waterproof DIN (conductivity sensor with built-in temperature)
Conductivity sensor (includes 013010A Orion DuraProbe 4-cell conductivity sensor)
1000 hours
IP-66
14° to 131°F (ambient)
16cm x 13.3cm x 3cm (6.30 in. x 5.24 in. x 1.18 in.)
0.6kg