Learn More
Thermo Scientific™ Orion™ 266S Advanced Waterproof Portable pH/mV Meter
Measure and log pH, mV and temperature using the Thermo Scientific™ Orion™ 266S Intrinsically Safe (US) Portable pH Meter
Manufacturer:  Thermo Scientific™ 0266S2
 View more versions of this product
Catalog No. 13-650-103
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Measure pH, mV and temperature in environments where non-sparking equipment is required using the Thermo Scientific™ Orion™ 266S Intrinsically Safe (US) Portable pH Meter. Log up to 100 data points and use the RS232 port to transfer data to an external device. Intrinsically safe approval rating by Factory Mutual (FM) for US only.
Features:
  • Meter can be controlled by a remote PC via RS-232-C output and software for transferring data.
  • Compact, portable waterproof meter simultaneously shows pH and temperature on watertight LCD.
  • Meter features mV mode, GLPcheck™ electrode verification and three-point autocalibration.
  • Three AA alkaline batteries supply power for up to 2000 hours of continuous use.
  • Meter can be controlled by a remote PC via RS-232-C output and software for transferring data to the computer.
Includes:
  • Meter Only package includes meter, built-in quiver, neckstrap and instruction manual, RS232 cable and software
  • Meter/Electrode package includes meter, Triode™ 3-in-1 Combination pH Electrode, built-in quiver, neckstrap and instruction manual
  • Meter Field Kit package includes meter; built-in quiver; neckstrap and instruction manual; case; stand; electrode holder; beakers; 4, 7, 10 buffers; RS232 cable and software
Compatible with:
0900A0 Orion Model 900A Printer
Description & Specifications
Specifications
Orion Intrinsically Safe 266S pH portable meter
Orion Intrinsically Safe 266S pH portable meter
-2.00 to 16.00pH
±0.01pH
0.1mV
pH
-20° to 120 °C, -4° to 248° F
±0.3°C
1
Measure, cal, print, scroll up, scroll down, STO, RCL, clock, power keys
Transfer single or all data points to printer or computer
Automatic by interval or event or manual data logging
E-DIN Waterproof (pH electrode), banana plug (ATC temperature probe)
pH electrode, ATC temperature probe (purchase separately)
1000 hours
IP-66
14° to 131°F (ambient)
16cm x 13.3cm x 3cm (6.30 in. x 5.24 in. x 1.18 in.)
0.6kg
3 year meter warranty
Orion Intrinsically Safe 266S pH/mV/Temperature Portable Meter
Orion 266S pH meter, 3 AA batteries
0.1, 0.01pH
±1300.0mV
0.1% of reading ±0.3 mV
1 to 3
0.1°C
Automatic with ATC temperature probe
LCD
pH with mV and temperature
100 with time and date stamp
English
RS232
3 AA batteries (included)
Three AA (included)
CE, FM (IS, Class I Div 1 Group A, B, C, D, T4), Ex (Intrinsically Safe)
-10° to 55°C (ambient)
1.4 lb.
None