Learn More
Thermo Scientific™ Orion™ 917001 Epoxy ATC Probe
Get fast, accurate measurements in lab or field with this ATC Probe, which automatically corrects pH measurements for variation in electrode slope due to temperature change.
Supplier:  Thermo Scientific™ 917001
Catalog No. 13-641-709A
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Get fast, accurate measurements in the lab or field. The Orion™ 917001 Epoxy ATC Probe automatically corrects pH measurements for variation in electrode slope due to temperature change.
Features:
  • Break-resistant
  • Suitable for rugged environment
  • Should not be used in organic solvents
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0 to 14
Epoxy Body
0°C to 80°C
Orion SA/EA series meters including the EA940 meter
12 cm, excluding cap
Store in air
±1°C
ATC Probe
0°C to 80°C
8 mm
1 m
Product Suggestions
Product Suggestions