Thermo Scientific™ Orion™ AQUAfast™ AQ4000 Colorimeter and Accessories Available on GSA/VA Contract for Federal Government customers only.

Measure common water and wastewater parameters using the Thermo Scientific™ Orion™ AQUAfast AQ4000 Colorimeter with 65 preprogrammed methods and Auto-Test ampoule compatibility

Manufacturer: Thermo Scientific™ AQ4000

 View more versions of this product

Catalog No. 13-642-157

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Description Colorimeter for use with 189 pre-programmed method
    Display Type Custom LCD
    Inputs Keypad 12 dual function keys with tactile feedback
    Measurement Ranges 0 to 2A
    Resolution ±2nm
    Built-in Timer Yes
    Humidity 90 at 50°C max. %
    Cuvette Details 13, 16, 24mm
    Incident Light Source Light Emitting Diode (LED)
    LED Wavelengths 420, 520, 580, 610nm
    Software Details Up to 190 and 10 user defined programmed methods
    Wavelength Selection Automatic
    Battery Life 2500 hr (Alkaline); 10000 hr (Lithium)
    Zero and Blank Yes
    Battery Type AA
    Data Results Output 100
    Operating Temperature Range 0° to 45°C
    Standby Mode Yes
    Waterproof Rating IP67
    Wavelength 420, 520, 580, 610nm

    Measure common colorimetric water and wastewater parameters using the Thermo Scientific™ Orion™ AQUAfast AQ4000 Colorimeter with 65 preprogrammed methods, automatic selection of four wavelengths and timer. Use Auto-Test ampoules with Auto-ID barcodes to simplify sample preparation and tests – ampoules contain pre-measured liquid reagent and fill with the exact volume of sample, colorimeter reads the barcode and automatically identifies the parameter and initiates the test. This IP67 waterproof meter records 100 data points and offers an average battery life of 2500 hours intermittent use.

    Features:

    • Identifies the species to be measured
    • Ensures error-free readings at a very affordable price
    • Selects the program and initiates the measurement without the user ever having pushed a button
    • Guides user through the setup and measurement with simple operation
    • Waterproof to IP67 standards
    Benefits:
    • User can customize calibration and create completely custom method.
    • Up to 12 data points can be used to define custom method.
    • User can store maximum of 10 methods in the colorimeter's memory.
    • User can datalog 100 points in the field with a time and date tag.

    Ordering Information:

    For use with Orion™ AQUAfast™ AQ4000, Orion™ AQUAfast™ AQ3070, Orion™ AQUAfast™ AQ3700 and discontinued Orion AQ2 series meters.

    Note: Uses Thermo Scientific™ Auto-ID™ ampoules, liquid, powder and tablet reagent including many methods accepted for EPA, ISO and other regulatory reporting.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit