Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents Available on GSA/VA Contract for Federal Government customers only.
Use Thermo Scientific™ Orion™ AQUAfast Auto-Test Ampoule Reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and testing with Auto-ID bar-coding
Manufacturer: Thermo Scientific™ AC4005
 View more versions of this product
Catalog No. 13-642-171
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    EPA Approved Methods Drinking Water: ASTM D3867-90 (B) / Standard Methods 4500-NO3- E; Wastewater: ASTM D3867-04 (B) / Standard Methods 4500-NO3- E
    For Use With Colorimetric water and wastewater testing equipment
    Recommended Accessories AQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER)
    Includes 30 Auto-ID Ampoules
    Item Description AQUAfast nitrate MR Auto-ID ampoules, 30 pack
    For Use With (Equipment) Orion AQUAfast AQ4000 colorimeter
    Quantity 30 ampoules
    Type Nitrate
    Test Method Cadmium Reduction
    Test Reagent Formats Ampoule
    Description AQUAfast nitrate MR Auto-ID ampoules, 30 pack
    Range 1.0 to 7.5mg/L N
    Use Thermo Scientific™ Orion™ AQUAfast Auto-Test Ampoule Reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The Auto-Test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
    • No. of test: 30
    • Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
    • Thermo Scientific™ Auto-ID™ eliminates measurement errors
  • Description & Specifications
Related Products
Product Content Feedback