Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ AQUAfast™ II Chemistries Test Kits

Use Thermo Scientific™ Orion™ AQUAfast Tablet Reagents to perform colorimetric measurements of common water parameters with convenient, rapidly dissolving tablets

Manufacturer:  Thermo Scientific™ AC2001

 View more versions of this product

Catalog No. 13-644-937


  Check Availability
Add to cart

Disclaimers:

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


Specifications

Description

Use Thermo Scientific™ Orion™ AQUAfast Tablet Reagents with Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf live accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.

  • For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ3070, Orion AQUAfast AQ3700 and discontinued Orion AQ2 series meters and othercolorimeters
  • No. of tests: 50 to 100
Description & Specifications

Specifications

Orion AquaMate 8000 UV-Vis Spectrophotometer, Orion AquaMate 7000 Vis Spectrophotometer, Orion AQUAfast AQ4000 colorimeter, Orion AQUAfast AQ3700 colorimeter
AQUAfast 24mm round vials (Cat. No. AC2V24)
Phenol Red
AQUAfast pH reagent tablets, 100 tests
Colorimetric water and wastewater testing equipment
AQUAfast pH reagent tablets, 100 tests
100 tests
pH
Tablet
6.5 to 8.4 pH
100 Phenol Red Tablets
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit