Learn More
Thermo Scientific™ Orion™ AQUAfast™ Reaction/Digestion Tubes and Liquid Reagents
Perform colorimetric testing including fluoride, COD, total nitrogen and total phosphate using the Thermo Scientific™ Orion™ AQUAfast Liquid Reagents, Digestion and Reaction Tubes.
$75.10 - $318.00
Specifications
For Use With (Equipment) Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast
View More Specs
Products 6
Catalog Number Mfr. No. Description Range Price Quantity & Availability  
Catalog Number Mfr. No. Description Range Price Quantity & Availability  
13-642-043
View Documents Promotion Details
Thermo Scientific™
CODH00
AQUAfast COD 0-1500 ppm digestion tubes, 25 pack 0 to 1500mg/L O2
Each for $75.30
Add to cart
13-642-044
View Documents Promotion Details
Thermo Scientific™
CODHP0
AQUAfast COD 0-15000 ppm digestion tubes, 25 pack 0 to 15000mg/L O2
Each for $75.80
Add to cart
13-642-042
View Documents Promotion Details
Thermo Scientific™
CODL00
AQUAfast COD 0-150 ppm digestion tubes, 25 pack 0 to 150mg/L O2
Each for $75.10
Add to cart
13-301-163
View Documents Promotion Details
Thermo Scientific™
CODH150
AQUAfast Digestion Tubes, Range: 0 to 15000 mg/L O2, COD High Range 0 to 15000 mg/L O2
Each for $311.50
Add to cart
13-301-159
View Documents Promotion Details
Thermo Scientific™
CODL150
AQUAfast COD 0-150 ppm digestion tubes, 150 pack 0 to 150 mg/L O2
Each for $311.50
Add to cart
13-301-162
View Documents Promotion Details
Thermo Scientific™
CODM150
AQUAfast COD 0-1500 ppm digestion tubes, 150 pack 0 to 1500 mg/L O2
Each for $318.00
Add to cart
Description
Description
Use Thermo Scientific™ Orion™ AQUAfast Digestion Tubes, Reaction Tubes and Liquid Reagents with Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers. Digestion and reaction tubes contain the precise amount of reagent, are made of special optical glass and fit the Orion™ AQUAfast COD165 thermoreactor. Digestion tubes measure COD, total nitrogen and total phosphate. Reaction tubes measure ammonia, nitrate and phosphate. Liquid reagents mix quickly and easily in solution and measure fluoride (SPADN), hydrazine and pH.
Changes in nutrient levels can alter the delicate balance in ecosystems and contaminate drinking water. Tube tests make it easy to determine concentrations of species in samples.
Specifications
Specifications
Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast
SDS
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.