Thermo Scientific™ Orion™ AquaPro™ pH Combination Electrodes Available on GSA/VA Contract for Federal Government customers only.

Read viscous liquid samples quickly and accurately without the sample coming into contact with silver ions by using these pH combination electrodes.

Manufacturer: Thermo Scientific™ 9103APWP

 View more versions of this product

Catalog No. 13-645-150

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Length (Metric) Cable 1m
    Connector Types Waterproof BNC
    Diameter (English) Body 0.26 in.
    pH Range 0 to 14
    Length (English) 6.5 in.
    Length (English) Cable 39 in.
    Temperature (English) Operating 32° to 140°F
    pH Relative Accuracy ±0.02pH
    Junction Open
    Length (Metric) 165mm
    Description Combination semi-micro pH electrode, Glass Body pH
    Diameter (Metric) Body 6.5mm
    Internal Reference Ag/AgCl Double Junction
    Applications Small sample sizes, test tubes
    Storage Requirements Always store in pH storage solution, Orion 910001. Either store in a beaker of storage solution or reassemble in the storage bottle the electrode was provided with.
    Temperature (Metric) Operating 0° to 60°C

    Read viscous liquid samples quickly and accurately without the sample coming into contact with silver ions with Thermo Scientific™ Orion™ AquaPro™ pH Combination Electrodes, which have a clog-resistant and open double junction reference system. These electrodes are ideal for measurements in samples containing proteins or TRIS buffers and that do not contain mercury. The electrodes require minimal maintenance because there is no fill solution; instead they have a polymer gel reference.

    Feature improved electrode life, low maintenance, fast response time and clean junctions. Patented reference system and double-junction design prevent silver ions from contacting sample.

    • Measure 0 to 14pH
    • Temperature range: 0° to 60°C (0° to 140°F)
    • Choice of connectors with 1m (3ft.) cable

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit