Learn More
Thermo Scientific™ Orion™ AutoTration™ 500 Autosampler Accessories
Maintain automated sample analysis with accessories for the legacy Thermo Scientific™ Orion™ AutoTration 500 Autosampler
$275.50 - $349.50
Specifications
Type Titrator Accessories
View More Specs
Products 2
Catalog Number Mfr. No. Container Size Price Quantity & Availability  
Catalog Number Mfr. No. Container Size Price Quantity & Availability  
13-641-964
View Documents
Thermo Scientific™
ASB050
50 mL beakers
Pack of 50 for $275.50
Add to cart
13-641-965
View Documents
Thermo Scientific™
ASB150
150 mL beakers
Pack of 50 for $349.50
Add to cart
Description
Description
Maintain automated sample analysis with accessories for the legacy Thermo Scientific™ Orion™ AutoTration 500 Autosampler.
  • Interchangeable racks to accommodate sample size and volume
  • Choice of 48- and 28-position racks
  • Beaker sizes of 45-, 50- and 150-mL
  • Replacement electrode holders for each model
Specifications
Specifications
Titrator Accessories
SDS
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.