Learn More
Thermo Scientific™ Orion™ AutoTration™ 500 Autosampler
Save time and enhance data with the Thermo Scientific™ Orion™ AutoTration™ 500 Autosampler.
Specifications
Certifications/Compliance CE
View More Specs
Products 1
Catalog Number Mfr. No. Container Size Price Quantity & Availability  
Catalog Number Mfr. No. Container Size Price Quantity & Availability  
13-644-908
View Documents
Thermo Scientific™
AT5150
150mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Save time and enhance data with the Thermo Scientific™ Orion™ AutoTration™ 500 Autosampler, which automatically performs analyses, calibrations and standardization procedures.
Efficient and durable:
  • Improves data quality by using up to five rinse stations
  • Constructed of durable, corrosion-resistant, sealed electronics chassis and coated metal parts to protect against harshest chemical environments
  • Compatible with Orion™ Star™ Plus meters and 960 Titrator PLUS for automated ISE techniques and titrations
  • RS-232 interface for easy communication
Includes:
AT5050: 48-position beaker rack for 50mL beakers; small electrode holder; drip tray; cable management system; BNC extension cable; stirrer extension cable; accessories package; interface cable; line cord; 50 pack of 50mL plastic beakers; instruction manual; warranty card.
AT5150: 28-position beaker rack for 150mL beakers; small electrode holder; drip tray; cable management system; BNC extension cable; stirrer extension cable; accessories package; interface cable; line cord; 50 pack of 150mL plastic beakers; instruction manual; warranty card.
Compatible with:
  • 1112000 - 3 Star Plus pH meter
  • 1114000 - 3 Star Plus conductivity meter
  • 1115000 - 4 Star Plus pH/ISE meter
  • 1117000 - 4 Star Plus pH/conductivity meter
  • 1119000 - 5 Star Plus pH/ISE/Conductivity/Dissolved Oxygen meter
  • ATB050 - Beakers, 50mL, 50/pk
  • ATB150 - Beakers, 150mL, 50/pk
  • 096S22 - Beakers, 45mL, 50/pk
Specifications
Specifications
CE