Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ Chlorine XP Water Quality Analyzer

Our Thermo Scientific™ Orion™ Chlorine XP is a DPD-based chlorine analyzer for reliably measuring free, total, and combined chlorine, and pH and temperature.

Manufacturer:  Thermo Scientific™ CXP71

 View more versions of this product

Catalog No. 13-642-100


  Check Availability
Add to cart

Specifications

Description

Perform routine water quality parameter measurements in your wastewater and drinking water applications with high accuracy and stability using the Thermo Scientific™ Orion™ Chlorine XP Water Quality Analyzer. With minimal maintenance, low cost of ownership, and multiple parameter measurement in one unit, our chlorine analyzer provides one of the most comprehensive water quality monitoring solutions on the market.

The Orion Chlorine XP Analyzer measures free, total, and combined chlorine as well as pH and temperature, providing accurate, objective, and instantaneous readings of chlorine in water. The colorimetric technology and auto calibration functionality minimizes the need for periodic calibrations and is compatible with all disinfectant systems. User-selectable reagent mixing allows users to increase usage per set, providing up to 60 days of unattended operation.

High Performance and Reliability

  • Proven accuracy and repeatability even in harsh sample conditions (sea water, colored water, oil water)
  • 0 to 10ppm measurement range – in most applications
  • Self zero calibration before each reading, enables "0" reading
  • Wet tested for 24 hours before shipping
  • 2-year warranty

Low Price and Cost of Ownership

  • Free and total chlorine, plus pH and temperature with optional ORP, conductivity, and turbidity functions in one analyzer – saves thousands of dollars over purchasing separate analyzers
  • Long reagent shelf life provides significant reagent savings compared to other manufacturers
Low-Maintenance Light Source Self-Calibration Adjustment
  • Hands-free self cleansing of the photocell
  • Automatic elimination of bubbling in the photocell
  • Maintenance reminders and alarms (once a year on average)
  • Auto buffer recognition at 4, 7, and 10
Description & Specifications

Specifications

Free Chlorine only with 2X 4/20mA outputs
±3%
+/-0.5%
100-115 VAC, 50/60Hz, 1.0A or 200-230 VAC 50/60Hz, 0.5A
33cm
67cm
14cm
11Kg
IP-65 rated enclosures (NEMA 4 equivalent)
0° to 100°C
2 Standard 4-20mA, 4 Optional 4-20mA
6 relays
-10° to 55°C
CSA, CE
Industrial free and total chlorine and pH measurements
1 inlet for water and 1 outlet for drainage
Chlorine
DPD
4-20mv
1 inlet
1 Stream
2 years
0.01ppm
2-10 minutes
13in
26in
5.5in
24lbs
Auto calibration
Up to 14.5 psi
Wall mount
CE and CSA certified
15° to 131°F
6 analog outputs
No Drift- zero adjustment before every reading
Basic setup
Up to 14.5 psi
0-10PPM Chlorine, 0-14 PPM pH
14.1 psi
Up to 14.5 psi
Up to 14.5 psi
26 x 5.5 x 13 in. (67 x 14 x 33cm)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit