Thermo Scientific™ Orion™ Chlorine XP Water Quality Analyzer

Our Thermo Scientific™ Orion™ Chlorine XP is a DPD-based chlorine analyzer for reliably measuring free, total, and combined chlorine, and pH and temperature.

Manufacturer: Thermo Scientific™ CXP71PH

 View more versions of this product

Catalog No. 13-642-102

  Check Availability
Add to Cart

    Specifications

    Warranty 2 years
    Resolution 0.01ppm
    Repeatability ±0.5%
    Inputs Chlorine
    Pressure 14.1 psi
    Height (English) 13in
    Weight (English) 24lbs
    Calibration Auto calibration
    Enclosure Rating IP-65 rated enclosures (NEMA 4 equivalent)
    Temperature (Metric) Operating -10° to 55°C
    Inlet Pressure Up to 14.5 psi
    Measurement Ranges 0-10PPM Chlorine, 0-14 PPM pH
    Response Time 2-10 minutes
    Length (English) 26in
    Certifications/Compliance CSA, CE
    For Use With Industrial free and total chlorine and pH measurements
    Inlet/Outlet 1 inlet for water and 1 outlet for drainage
    Sample Flow Rate Up to 14.5 psi
    Sample Pressure Up to 14.5 psi
    Sample Streams 1 Stream
    Length (Metric) 67cm
    Accuracy ±3%
    Relay Details 6 relays
    Temperature (English) Operating 15° to 131°F
    Methods DPD
    Range (mV) 4-20mv
    Voltage 100-115 VAC, 50/60Hz, 1.0A or 200-230 VAC 50/60Hz, 0.5A
    Height (Metric) 33cm
    Weight (Metric) 11Kg
    Sample Temperature 0° to 100°C
    Mounting Options Wall mount
    Outputs 2 Standard 4-20mA, 4 Optional 4-20mA
    Dimensions (L x W x H) 26 x 5.5 x 13 in. (67 x 14 x 33cm)
    Description Free Chlorine + pH + Temp. 2x4/20mA outputs
    Width (English) 5.5in
    Width (Metric) 14cm
    Sample Inlet 1 inlet
    Flow Rate Up to 14.5 psi
    Regulatory Details CE and CSA certified
    Analog Outputs 6 analog outputs
    Drift No Drift- zero adjustment before every reading

    Perform routine water quality parameter measurements in your wastewater and drinking water applications with high accuracy and stability using the Thermo Scientific™ Orion™ Chlorine XP Water Quality Analyzer. With minimal maintenance, low cost of ownership, and multiple parameter measurement in one unit, our chlorine analyzer provides one of the most comprehensive water quality monitoring solutions on the market.

    The Orion Chlorine XP Analyzer measures free, total, and combined chlorine as well as pH and temperature, providing accurate, objective, and instantaneous readings of chlorine in water. The colorimetric technology and auto calibration functionality minimizes the need for periodic calibrations and is compatible with all disinfectant systems. User-selectable reagent mixing allows users to increase usage per set, providing up to 60 days of unattended operation.


    High Performance and Reliability

    • Proven accuracy and repeatability even in harsh sample conditions (sea water, colored water, oil water)
    • 0 to 10ppm measurement range – in most applications
    • Self zero calibration before each reading, enables "0" reading
    • Wet tested for 24 hours before shipping
    • 2-year warranty


    Low Price and Cost of Ownership

    • Free and total chlorine, plus pH and temperature with optional ORP, conductivity, and turbidity functions in one analyzer – saves thousands of dollars over purchasing separate analyzers
    • Long reagent shelf life provides significant reagent savings compared to other manufacturers

    Low-Maintenance Light Source Self-Calibration Adjustment

    • Hands-free self cleansing of the photocell
    • Automatic elimination of bubbling in the photocell
    • Maintenance reminders and alarms (once a year on average)
    • Auto buffer recognition at 4, 7, and 10

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit