Learn More
Thermo Scientific™ Orion™ Cyanide Electrodes Available on GSA/VA Contract for Federal Government customers only.
Measure cyanide ions in aqueous solutions quickly, simply, accurately and economically with the Thermo Scientific™ Orion™ Cyanide Electrodes.
Manufacturer:  Thermo Scientific™ 9406SC
 View more versions of this product
Catalog No. 13-642-570
  Check Availability
Add to cart
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Measure cyanide ions in aqueous solutions quickly, simply and economically with Thermo Scientific™ Orion™ Cyanide Electrodes.
No separate reference electrode is needed with this electrode, making it convenient to use with small sample volumes.
  • Calibration Standards Catalog number: Consult instruction manual
  • Required ISA Catalog number: 951011
Description & Specifications
Specifications
Combination Ion Selective Electrode
With screw cap connector
ISE Meters
Screw Cap
Free-flowing liquid junction assures stable, drift-free potentials