Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ Dual Star™ pH, ISE, mV, ORP and Temperature Dual Channel Benchtop Meter PROMO

Accurately monitor pH, ion concentration, mV, ORP and temperature on two channels with the Thermo Scientific™ Orion™ Dual Star pH / ISE Benchtop Meter for advanced lab analysis.

Specifications

Accuracy (mV) ±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (Temperature) ±0.1°C
Calibration pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
Certifications/Compliance CE, TUV 3-1, FCC Class A
Calibration Points (ISE) 2 to 6
View More Specs

Products 2
Catalog Number Mfr. No. Description Includes Price Quantity & Availability  
Catalog Number Mfr. No. Description Includes Price Quantity & Availability  
13-642-781SD
View Documents
Thermo Scientific™
2115101SD
Orion Dual Star pH/ISE dual channel benchtop meter with ROSS Ultra glass-body pH electrode, ATC probe and stand Dual Star pH/ISE dual channel meter, 8102BNUWP ROSS Ultra glass-body pH electrode, 927007MD Orion stainless steel ATC probe, electrode stand, universal power adapter, literature, computer interface cable and meter test certificate This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
13-642-782SD
View Documents
Thermo Scientific™
2115001SD
Orion Dual Star pH/ISE dual channel benchtop meter kit with ROSS Ultra glass-body pH electrode, ATC probe, solutions and stand Dual Star Meter with Electrode Stand; Universal Power Adapter; 8102BNUWP ROSS Ultra Glass Refillable pH Electrode; 927007MD Stainless Steel ATC Probe; 810001 ROSS Storage Solution; 910104, 910107, 910110 pH 4.01, 7.00, 10.01 pH Buffers This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

The Orion Dual Star pH/ISE dual channel benchtop meter accelerates sample measurement throughput by showing the results of two channels simultaneously on its large, backlit display. Measure pH, ion concentration (ISE), mV, relative mV or ORP with temperature on two channels using the meter’s two BNC, reference pin-tip and ATC temperature inputs. Collect critical measurements using an assortment of read modes and onscreen ready indicator that alerts when readings are stable. The meter’s Smart Stability and Smart Averaging functions automatically compensate for measurement conditions, so the meter displays more accurate results faster.
 

  • View large individual measurements on a full screen display or quickly toggle to a split screen display to view two measurements at the same time
  • Easy-to-follow onscreen text prompts for calibration and setup menus available in a multi-language user interface with English, Spanish, French, German or Italian options
  • EZ Startup menu facilitates quick and easy first time meter setup
  • Numeric keypad and menu-specific function keys allow for quick and efficient meter setup and data entry
  • Selectable read modes include Auto-Read, On-Ready, At Timed Intervals, Continuous, Value Change in Measurement and Timed for a variety of measurement collection methods for different analytical methods
  • Perform up to a 6 point pH calibration with automatic recognition for USA/NIST and EURO/DIN buffers with option to manually input custom buffer values
  • Perform up to a 6 point ion concentration (ISE) calibration with non-linear auto-blank and low-concentration stability options or use incremental methods for single/double known addition and single/double known subtraction
  • Non-volatile meter memory holds up to 1000 data points with time and date stamp
  • Easily transfer data via USB and RS232 to a printer or computer with free Orion Dual Star Com communication software
  • Stir samples with ease using the meter-controlled Orion Star stirrer probe
  • Meter-attachable electrode stand assists with moving and maintaining electrodes
  • IP54-rated housing handles splashes and is wall-mountable to save space

 

Warranty: Three-year meter replacement warranty

 

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications

Specifications

±0.2 mV or ±0.05% of reading whichever is greater
pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
2 to 6
0.0001 to 19,900 in ppm, M, mg/L, % or unit-less
0.0001 minimum, 1 to 3 significant digits (user selectable)
Graphic LCD with Backlight
Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
±0.1°C
CE, TUV 3-1, FCC Class A
1 to 6
-2.000 to 19.999
-2.000 to 19.999
Universal 50 to 60 Hz, 100 to 240 VAC power adapter (Cat. No. 1010003)
USB, RS232
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit