Learn More
Thermo Scientific™ Orion™ Electrode Adapter Cables Available on GSA/VA Contract for Federal Government customers only.
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
Manufacturer:  Thermo Scientific™ 090035
 View more versions of this product
Catalog No. 13-301-64
  Check Availability
Add to cart
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
  • For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2mm reference pins or just a stripped lead inputs
  • Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors
Description & Specifications
Specifications
Meters with 2mm reference pins connectors
LEMO Miniature (Mettler); BNC; BNC to LEMO Miniature Adapter
Electrode Adapter Cables
BNC
LEMO Miniature (Mettler)