Thermo Scientific™ Orion™ Electrode Adapter Cables
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
Manufacturer: Thermo Scientific™ 91USCB
 View more versions of this product
Catalog No. 13-641-46
  N/A
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Connector Types Combination Electrode with Screw Cap
    Description U.S. Standard; Combination Electrode with Screw Cap; Detachable Cable to U.S. Standard Connector
    Meter Input U.S. Standard
    Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
    • For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2mm reference pins or just a stripped lead inputs
    • Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors
  • Description & Specifications
Related Products
Product Content Feedback