Thermo Scientific™ Orion™ Electrode Adapter Cables Available on GSA/VA Contract for Federal Government customers only.
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
Manufacturer: Thermo Scientific™ 090033
 View more versions of this product
Catalog No. 13-641-712A
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Description BNC; U.S. Standard; U.S. Standard to BNC Adapter
    Connector Types U.S. Standard
    Meter Input BNC
    Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
    • For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2mm reference pins or just a stripped lead inputs
    • Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors
  • Description & Specifications
Related Products
Product Content Feedback