Thermo Scientific™ Orion™ Electrode Adapter Cables
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
Manufacturer: Thermo Scientific™ 91CDIN
 View more versions of this product
Catalog No. 13-641-50
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Connector Types Screw Cap
    Item Description E DIN (Knick, Schott, WTW); Screw Cap; Detachable Cable to E Din Connector
    Description E DIN (Knick, Schott, WTW); Screw Cap; Detachable Cable to E Din Connector
    Meter Input E DIN (Knick, Schott, WTW)
    Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
    • For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2mm reference pins or just a stripped lead inputs
    • Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors
  • Description & Specifications
Related Products
Product Content Feedback