Learn More
Thermo Scientific™ Orion™ Electrode Output Adapters
Use Thermo Scientific? Orion? Electrode Output Adapters with a variety of meter inputs.
Manufacturer:  Thermo Scientific™ 1010050-WA
 View more versions of this product
Catalog No. 13-645-131
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Use Thermo Scientific™ Orion™ Electrode Output Adapters with a variety of meter inputs.
Description & Specifications
Specifications
310, 320, 330, 350, 370, 555A, 550A, 535A, 162A
MiniDIN ATC Probe to 3.5mm Phono Tip meter input