Learn More
Thermo Scientific™ Orion™ Electrode Output Adapters
Use Thermo Scientific? Orion? Electrode Output Adapters with a variety of meter inputs.
Manufacturer:  Thermo Scientific™ 1010901
 View more versions of this product
Catalog No. 13-645-136
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Use Thermo Scientific™ Orion™ Electrode Output Adapters with a variety of meter inputs.
Description & Specifications
Specifications
50Aplus, 145Aplus, 125Aplus, 115Aplus, 105Aplus
MiniDIN Conductivity Probe connector to 8-Pin DIN Conductivity meter input