Thermo Scientific™ Orion™ Green pH Combination Electrodes Available on GSA/VA Contract for Federal Government customers only.

Go environmentally friendly by choosing one of the first lab pH electrodes to meet RoHS requirements.

Manufacturer: Thermo Scientific™ GS9156BNWP

 View more versions of this product

Catalog No. 13-620-152

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Length (Metric) Cable 1m
    Connector Types Waterproof BNC connector
    Material Durable epoxy body
    Junction Single
    Description Liquid-filled
    pH Range 0 to 14
    Length (English) Cable 39.3 in.

    Go environmentally friendly by choosing the Thermo Scientific™ Orion™ Green pH Combination Electrode. It is one of the first lab pH electrodes to meet RoHS requirements by containing no lead, mercury or other hazardous substances — making disposal of old electrodes easy and hassle free.

    • No disposal issues, no disposal cost
    • Choice of single-junction models for clean-water measurements, or double-junction models for dirty-water testing
    • Choice of refillable or low-maintenance gel-filled models
    • Duarable epoxy body
    • pH Range: 0 to 14
    • Dimensions: 120 x 12mm, cap dia. 16mm
    • Waterproof BNC connector, 1m cable; fit any meter with BNC input

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit