Thermo Scientific™ Orion™ Iodide Electrodes
Get fast and stable readings using Thermo Scientific™ Orion™ Iodide Electrodes.
Manufacturer: Thermo Scientific™ 9453SC
 View more versions of this product
Catalog No. 13-641-33
  N/A
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Connector Types Screw cap
    Description With screw cap connector
    Get fast and stable readings using Thermo Scientific™ Orion™ Iodide Electrodes with built-in Thermo Scientific™ Sure-Flow™ reference. Combination design decreases the amount of required sample and reduces waste.
    • Calibration standards catalog number: 945306 - 0.1M NaI Required ISA catalog number: 940011
  • Description & Specifications
Related Products
Product Content Feedback