Thermo Scientific™ Orion™ Metallic Combination Electrodes Available on GSA/VA Contract for Federal Government customers only.
Perform accurate Karl Fischer titrations with double platinum Thermo Scientific™ Orion™ Metallic Combination Electrodes.
Manufacturer: Thermo Scientific™ 977900
 View more versions of this product
Catalog No. 13-299-109
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Description Double platinum KF glass electrode
    For Use With KF coulometric titrations
    Temperature (English) Operating 32° to 104°F
    Item Description Model 977900; Double platinum KF glass electrode
    Temperature (Metric) Operating 0° to 40°C
    Material Dual platinum
    Perform accurate Karl Fischer titrations with double platinum Thermo Scientific™ Orion™ Metallic Combination Electrodes.
  • Description & Specifications
Related Products
Product Content Feedback