Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ pH Electrode Cleaning and Storage Solutions

Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Cleaning and Storage Solutions.

Manufacturer:  Thermo Scientific™ 9100CB

 View more versions of this product

Catalog No. 13-301-68


Add to cart

Description

Description

Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Cleaning and Storage Solutions in ready to use bottles.

Cleaning Solutions

  • Solutions are identified by the letters A, B, C, and D, each letter designates a unique formulation
  • Cleaning solution A is for removing protein contaminants, cleaning solution B is for removing bacterial contaminants, cleaning solution C is for general cleaning and cleaning solution D is for removing oil and grease contaminants
  • Bottles of the ready-to-use cleaning solutions include a small beaker to hold the cleaning solution and a plastic pipet to remove the old electrode filling solution
  • pH Electrode Cleaning Solution Kit (900020) includes one bottle each of cleaning solutions A, B, C and D; one beaker and one pipet

Storage Solutions

  • Storage solution ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples

Note:

The following part numbers have been changed due to a recent system upgrade.

Old Part Number: 900021

New Part Number: 900021-WA

Old Part Number: 900022

New Part Number: 900022-WA

Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Data Safety Sheet (MSDS) files.

Specifications

Specifications

pH Electrode Storage Solution, 5 gallon (19L) Cubitainer
pH Electrode
19 L
Ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples
Cubitainer
pH Electrode Storage Solution
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.