Learn More
Thermo Scientific™ Orion™ Protective Guards for Conductivity Cells and Dissolved Oxygen Probes
Provide additional protection and weight with Thermo Scientific™ Orion™ Protective Guards for Conductivity Cells and Dissolved Oxygen Probes.
$157.30
Products 1
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
13-642-156
View Documents
Thermo Scientific™
081045
Plastic
Each for $157.30
Add to cart
Description
Description
Provide additional protection and weight for conductivity cells and DO probes with Thermo Scientific™ Orion™ Protective Guards for Conductivity Cells and Dissolved Oxygen Probes. Use the all stainless-steel guard when optimum protection and weight are required. Combine the stainless steel cap and plastic-sleeve when weight is needed for submersion and standard protection is needed. Or, when minimal weight and protection are required, use the all-plastic guard.
For Use With
  • Conductivity Cells: 013005MD, 013010MD, 0130020MD, 013025MD, 013005A, 013010A, and 013005D
  • Dissolved Oxygen Probes: 080510MD, 0830005MD, 083010MD, 083025MD, 083060MD, 083005A, 083010A, and 083005D
Warranty: Out of box
SDS
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.