Learn More
Thermo Scientific™ Orion™ Resistor Kits and Flow Cell Conductivity Probe Accessories
Description
Confidently perform conductivity tests and measurements with Thermo Scientific™ Orion™ Accessories. Find an additional or replacement Glass Flow Cell for use with the Orion 2-Cell Ultra-pure Conductivity Probe. Use the Thermo Scientific Orion Conductivity Resistor Calibration Kit to verify the performance of Thermo Scientific™ Orion™ Versa Star Pro™ and Orion Star™ Bench and Portable Conductivity Meters. Each conductivity calibration kit includes six resistors that simulate nominal conductance values between 1 μS and 100 mS and have 8 pin MiniDIN connectors.
Conductivity Standards
- Traceable to NIST standard reference materials
- Designed to meet all end user requirements, including laboratory grade and DIN standards
- Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature
Specifications
Specifications
| Description | Conductivity Calibration Resistor Kit with 8 Pin MiniDIN Connection |
| For Use With (Equipment) | 3-Star Conductivity Meter, 4-Star pH/Conductivity Meter, 5-Star pH/DO/Conductivity Meter, 5-Star pH/ISE/DO/Conductivity Meter |
| Type | Calibration Resistor Kit |
| For Use With (Application) | Conductivity Calibration |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.