Learn More
Thermo Scientific™ Orion™ Resistor Kits and Accessories
Use the Thermo Scientific™ Orion™ Conductivity Resistor Kit to verify the performance of Versa Star and Star A series conductivity meters with 8 pin MiniDIN connectors
$261.50 - $516.00
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
09-357-44
View Documents
Thermo Scientific™
013017
Replacement Flow Cell Each for $261.50
Add to cart
13-303-154
View Documents
Thermo Scientific™
1010001
Conductivity Calibration Resistor Kit with 8 Pin MiniDIN Connection Each for $516.00
Add to cart
Description
Description
Use the Thermo Scientific™ Orion™ Conductivity Resistor Calibration Kit to verify the performance of Thermo Scientific™ Orion™ Versa Star conductivity meters, Thermo Scientific™ Orion™ Star A series conductivity meters and legacy Orion™ Star series conductivity meters. Each conductivity calibration kit includes six resistors that simulate nominal conductance values between 1 µS and 100 mS and have 8 pin MiniDIN connectors.
Conductivity Standards
  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards
  • Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.