Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ ROSS™ Combination Rugged Bulb pH Electrodes PROMO

When you purchase a Thermo Scientific™ ROSS™ pH Electrode, you’ll receive a ROSS Buffer Kit at no additional cost. Order using the catalog numbers below and your products will ship together.

$515.50 - $534.50

Specifications

Drift <0.002 pH/day
Filling Solution Type 3M KCl
Internal Reference ROSS
Junction Ceramic
Material Glass Body
View More Specs

Products 2
Catalog Number Mfr. No. Connector Types For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Connector Types For Use With (Equipment) Price Quantity & Availability  
13-641-805PMO
View Documents
Thermo Scientific™
8104BNKIT
BNC Standard pH Meter, BNC connector
Each for $534.50
Add to cart
 
13-641-4PMO
View Documents
Thermo Scientific™
8104SCKIT
Screw Cap Standard pH Meter, with separate cable with screw cap connector
Each for $515.50
Add to cart
 
Description

Description

The 8104BN Ross Combination Rugged Bulb pH Electrode works in extreme cases of samples varying from one another by 50°C or more with drift less than 0.002 pH per day and minimized restandardization.

  • Material: glass body
  • pH range: 0 to 14
  • Temperature range: 0 to 100°C
  • Internal reference: ROSS
  • Junction: ceramic
  • Dimensions: 120 × 12 mm
  • pH precision: 0.01
  • Fill solution: 810007
  • Cap diameter: 16 mm
Specifications

Specifications

<0.002 pH/day
ROSS
Glass Body
3M KCl
Ceramic
0 to 14
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit