Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes for Orion Series Meters

Measure biological media, pharmaceuticals, foods and more with Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes.

Manufacturer:  Thermo Scientific™ 8157UWMMD

 View more versions of this product

Catalog No. 13-643-202

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Measure biological media, pharmaceuticals, foods and more with Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes. They are ideal for any application where the use of metallic reference electrodes would contaminate the samples. ROSS Ultra pH/ATC electrodes offer more flexibility, accuracy and precision for your pH measurement needs. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements. They have great stability and drift and come with a two-year replacement warranty.

Unique internal element system replaces Ag/AgCl or Hg2Cl2 internals with platinum wire and redox-couple filling solution. Double-junction reference cells have porous ceramic or glass fiber outer junction. Design provides stable potential, independent of temperature, and stable readings even when samples vary by 50°C (90°F) or more. Minimizes drift, stirring effects, clogged junctions; no need for frequent standardization so samples are analyzed faster.

  • Epoxy body for durability and ruggedness
  • Built-in ATC probe
  • General-purpose applications
  • Standard size and standard bulb
  • Glass fiber junction
  • L x Dia.: 120 x 12mm

Specifications

  • 0 to 14pH
  • 0° to 100°C (32° to 212°F) range
  • Slope, 92 to 102%; isopotential point, pH 7
  • When standardized in 6.86pH buffer at 25°C (77°F), accuracy is ±0.03pH for the buffer from 0° to 100°C (32° to 212°F) (using ATC); response is stable to 0.01pH within 30 seconds at 25°C (77°F) and 75°C (167°F) and stable to ±0.002pH within 15 seconds when cycling between pH 6.86 and 4.01 buffers at 25°C (77°F)
  • Max. cap dia. (top): 0.75in. (19mm); collar dia. 0.62in. (16mm); Cable length is 39.37in. (1m)
Description & Specifications

Specifications

Orion ROSS Ultra Refillable pH/ATC Triode Combination Electrodes with a BNC and 8-pin MiniDIN Connector and 3 meter cable
2 years
0.01
0° to 100°C
Glass fiber
ROSS electrode 3 M KCl fill solution (Cat. No. 810007)
12mm
BNC Waterproof, 8-pin MiniDIN
ROSS
Orion Versa Star, Star A and Dual Star Meters
0 to 14
0.03pH for buffers using automatic temperature compensation
Rugged epoxy body
7
120mm
3m
Thermo Scientific Orion Versa Star, Star A, and Dual Star meters
pH/ATC Combination Electrode
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit