Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes PROMO

When you purchase a Thermo Scientific™ ROSS™ pH Electrode, you’ll receive a ROSS Buffer Kit at no additional cost. Order using the catalog numbers below and your products will ship together.

$390.00 - $648.00

Specifications

Filling Solution Type ROSS electrode 3 M KCl fill solution (Cat. No. 810007)
Internal Reference ROSS
Junction Glass Fiber
Material Epoxy Body
pH Range 0 to 14
View More Specs

Products 2
Catalog Number Mfr. No. Connector Types For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Connector Types For Use With (Equipment) Price Quantity & Availability  
13-643-201PMO
View Documents
Thermo Scientific™
8157BNUMDKIT
BNC & 8-pin MiniDIN Orion Versa Star, Star A and Dual Star Meters
Each for $648.00
Add to cart
 
13-642-676PMO
View Documents
Thermo Scientific™
8157BNURCAKIT
BNC/RCA Mettler Toledo Five and Seven series pH meters
Each for $390.00
Add to cart
 
Description

Description

Unique internal element system replaces Ag/AgCl or Hg2Cl2 internals with platinum wire and redox-couple filling solution. Double-junction reference cells have porous ceramic or glass fiber outer junction. Design provides stable potential, independent of temperature, and stable readings even when samples vary by 50°C (90°F) or more. Minimizes drift, stirring effects, clogged junctions; no need for frequent standardization so samples are analyzed faster.

  • Epoxy body for durability and ruggedness
  • Built-in ATC probe
  • General-purpose applications
  • Standard size and standard bulb
  • Glass fiber junction
  • L × Dia.: 120 × 12 mm

Specifications

  • 0 to 14 pH
  • 0 to 100°C (32 to 212°F) range
  • Slope, 92 to 102%; isopotential point, pH 7
  • When standardized in 6.86 pH buffer at 25°C (77°F), accuracy is ±0.03 pH for the buffer from 0 to 100°C (32 to 212°F) (using ATC); response is stable to 0.01 pH within 30 seconds at 25°C (77°F) and 75°C (167°F) and stable to ±0.002 pH within 15 seconds when cycling between pH 6.86 and 4.01 buffers at 25°C (77°F)
  • Max. cap dia. (top): 0.75 in. (19 mm); collar dia. 0.62 in. (16 mm); Cable length is 39.37 in. (1 m)
Specifications

Specifications

ROSS electrode 3 M KCl fill solution (Cat. No. 810007)
Glass Fiber
0 to 14
ROSS
Epoxy Body
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit