Thermo Scientific™ Orion™ Redox/ORP/Temp Electrodes Available on GSA/VA Contract for Federal Government customers only.
Efficiently measure ORP and temperature simultaneously without a separate reference electrode in a single instance.
Manufacturer: Thermo Scientific™ 9180BN
 View more versions of this product
Catalog No. 13-640-1
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    For Use With (Equipment) A+ Series Meters
    Material Epoxy
    Description Epoxy Refillable Triode
    Efficiently measure ORP and temperature simultaneously without a separate reference electrode in a single instance. Thermo Scientific™ Orion™ Redox/ORP/Temp Electrodes measure ORP and temperature using a platinum redox sensor, a silver/silver chloride internal reference system and a built-in thermistor which is adapted for Automatic Temperature Compensation (ATC).
    Features:
    • Available in epoxy triode low maintenance and epoxy refillable triode construction
    • Built-in thermistor for Automatic Temperature Compensation (ATC)
    • Not designed for amperometric or constant current titrations, such as Karl Fischer analysis
  • Description & Specifications
Related Products
Product Content Feedback