Learn More
Thermo Scientific™ Orion™ Two-Electrode Conductivity Cells Available on GSA/VA Contract for Federal Government customers only.
Perform applications in either the lab or the field with these versatile two-electrode conductivity cells, available in a variety of materials and measurement ranges.
Manufacturer:  Thermo Scientific™ 011010A
 View more versions of this product
Catalog No. 13-302-3
  Check Availability
Add to cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Perform applications in either the lab or the field with versatile Thermo Scientific™ Orion™ Two-Electrode Conductivity Cells, available in a variety of materials and measurement ranges.
2-Electrode conductivity cells 011010A and 011010 are glass/platinum, have a nominal cell constant of 1.0cm-1 and are intended for laboratory use; measurement range is 1µS/cm to 200mS/cm.
2-Electrode conductivity cells 011020A, 011020 and 9902BN are glass/platinum, have a nominal cell constant of 0.1cm-1 and are intended for laboratory use and ultra pure water; measurement range is 0.1µS/cm to 100µS/cm.
2-Electrode conductivity cells 011050MD, 011050A and 011050 are epoxy/platinum, have a nominal cell constant of 1.0cm-1 and are intended for field or laboratory use; measurement range is 1µS/cm to 20mS/cm.
2-Electrode conductivity cells 011510MD and 011510are epoxy/graphite, have a nominal cell constant of 1.0cm-1 and are intended for field or laboratory use; measurement range is 10µS/cm to 200mS/cm.
2-Electrode conductivity cells 013016MD, 013016A and 013016D are steel (V4A), have a nominal cell constant of 0.1cm-1 and are intended for laboratory use and ultra pure water; measurement range is 0.01µS/cm to 300µS/cm
2-Electrode conductivity cell 018020MD is glass/platinum, has a nominal cell constant of 10cm-1 and is intended for high electrolyte concentration solutions; measurement range is 10µS/cm to 2000mS/cm.
Recommended for:
Intended for laboratory and field use
Description & Specifications
Specifications
Cable: 8 pin Waterproof DIN
555A, 550A, 162A, 136A, 135A, 131S, 130A meters
1µS/cm to 200mS/cm
2-Electrode Conductivity Cell
Glass/Platinum, platinized